Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118946 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTFR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNTFR - ciliary neurotrophic factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207011.1 | Intron | NP_001193940.1 | ||||
NM_001842.4 | Intron | NP_001833.1 | ||||
NM_147164.2 | Intron | NP_671693.1 | ||||
XM_017014260.1 | Intron | XP_016869749.1 | ||||
XM_017014261.1 | Intron | XP_016869750.1 | ||||
XM_017014262.1 | Intron | XP_016869751.1 | ||||
XM_017014263.1 | Intron | XP_016869752.1 | ||||
XM_017014264.1 | Intron | XP_016869753.1 | ||||
XM_017014265.1 | Intron | XP_016869754.1 |
Set Membership: |
HapMap |