Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHPP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LHPP - phospholysine phosphohistidine inorganic pyrophosphate phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167880.1 | Intron | NP_001161352.1 | ||||
NM_001318331.1 | Intron | NP_001305260.1 | ||||
NM_001318332.1 | Intron | NP_001305261.1 | ||||
NM_022126.3 | Intron | NP_071409.3 | ||||
XM_005270026.3 | Intron | XP_005270083.1 | ||||
XM_006717943.1 | Intron | XP_006718006.1 | ||||
XM_011540058.2 | Intron | XP_011538360.1 | ||||
XM_017016509.1 | Intron | XP_016871998.1 | ||||
XM_017016510.1 | Intron | XP_016871999.1 | ||||
XM_017016511.1 | Intron | XP_016872000.1 | ||||
XM_017016512.1 | Intron | XP_016872001.1 |
Set Membership: |
HapMap |