Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGATGTGTGAGTGGACTAGTTT[C/G]TTTCAAGTCACACCATTCTTCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986791 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107986791 - uncharacterized LOC107986791 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2J4 - RNA polymerase II subunit J4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2D4 - ubiquitin conjugating enzyme E2 D4 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015983.3 | Intron | NP_057067.1 | ||||
XM_006715734.3 | Intron | XP_006715797.1 | ||||
XM_011515420.2 | Intron | XP_011513722.1 | ||||
XM_011515422.2 | Intron | XP_011513724.1 | ||||
XM_011515423.2 | Intron | XP_011513725.1 | ||||
XM_017012292.1 | Intron | XP_016867781.1 | ||||
XM_017012293.1 | Intron | XP_016867782.1 | ||||
XM_017012294.1 | Intron | XP_016867783.1 | ||||
XM_017012295.1 | Intron | XP_016867784.1 |
Set Membership: |
HapMap |