Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601332 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MKX - mohawk homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242702.1 | 1550 | UTR 3 | NP_001229631.1 | |||
NM_173576.2 | 1550 | UTR 3 | NP_775847.2 | |||
XM_017016105.1 | 1550 | UTR 3 | XP_016871594.1 | |||
XM_017016106.1 | 1550 | UTR 3 | XP_016871595.1 | |||
XM_017016107.1 | 1550 | Intron | XP_016871596.1 | |||
XM_017016108.1 | 1550 | UTR 3 | XP_016871597.1 |
Set Membership: |
HapMap |