Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGGGAATTGAGGACCCTTACT[C/T]ACCTTAGGCAGCGTCCTTGTTACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103740 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADH4 - alcohol dehydrogenase 4 (class II), pi polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000670.4 | 1340 | Intron | NP_000661.2 | |||
NM_001306171.1 | 1340 | Intron | NP_001293100.1 | |||
NM_001306172.1 | 1340 | Intron | NP_001293101.1 | |||
XM_017007713.1 | 1340 | UTR 3 | XP_016863202.1 | |||
XM_017007714.1 | 1340 | UTR 3 | XP_016863203.1 | |||
XM_017007715.1 | 1340 | UTR 3 | XP_016863204.1 |
LOC100507053 - uncharacterized LOC100507053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |