Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGCCTGTCCCACACCCTCTTC[C/T]ATCATGGCCTGGGCTAGTCTTTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606119 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100288181 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100288181 - uncharacterized LOC100288181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLURP1 - secreted LY6/PLAUR domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THEM6 - thioesterase superfamily member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016647.2 | Intron | NP_057731.1 | ||||
XM_005250955.2 | Intron | XP_005251012.1 |