Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603515 MIM: 139250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSHL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSHL1 - chorionic somatomammotropin hormone like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318.3 | 375 | Missense Mutation | GAA,GAC | E,D 47 | NP_001309.3 | |
NM_001321067.1 | 375 | Missense Mutation | GAA,GAC | E,D 81 | NP_001307996.1 | |
NM_001321068.1 | 375 | Missense Mutation | GAA,GAC | E,D 79 | NP_001307997.1 | |
NM_001321069.1 | 375 | Missense Mutation | GAA,GAC | E,D 102 | NP_001307998.1 | |
NM_022579.2 | 375 | Missense Mutation | GAA,GAC | E,D 141 | NP_072101.1 | |
NM_022580.2 | 375 | Missense Mutation | GAA,GAC | E,D 58 | NP_072102.1 | |
NM_022581.2 | 375 | Missense Mutation | GAA,GAC | E,D 118 | NP_072103.1 | |
XM_011524344.1 | 375 | Missense Mutation | GAA,GAC | E,D 141 | XP_011522646.1 | |
XM_011524346.2 | 375 | Missense Mutation | GAA,GAC | E,D 81 | XP_011522648.1 |
GH1 - growth hormone 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |