Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCCACTCAGGACTCACCTGCT[A/G]CTCACACCAGCCGCCTCCGCTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607212 MIM: 602777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD9 - caspase recruitment domain family member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNAPC4 - small nuclear RNA activating complex polypeptide 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266096.2 | 5425 | UTR 3 | XP_005266153.1 | |||
XM_006717241.2 | 5425 | UTR 3 | XP_006717304.1 | |||
XM_006717242.3 | 5425 | UTR 3 | XP_006717305.1 | |||
XM_006717244.2 | 5425 | UTR 3 | XP_006717307.1 | |||
XM_017015058.1 | 5425 | UTR 3 | XP_016870547.1 |