Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606615 MIM: 602140 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIF1AN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIF1AN - hypoxia inducible factor 1 alpha subunit inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017902.2 | 147 | Missense Mutation | CCC,GCC | P,A 41 | NP_060372.2 | |
XM_011539940.2 | 147 | Missense Mutation | CCC,GCC | P,A 41 | XP_011538242.1 | |
XM_011539941.1 | 147 | Intron | XP_011538243.1 |
NDUFB8 - NADH:ubiquinone oxidoreductase subunit B8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |