Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607423 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
POMT1 - protein O-mannosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077365.1 | Intron | NP_001070833.1 | ||||
NM_001077366.1 | Intron | NP_001070834.1 | ||||
NM_001136113.1 | Intron | NP_001129585.1 | ||||
NM_001136114.1 | Intron | NP_001129586.1 | ||||
NM_007171.3 | Intron | NP_009102.3 | ||||
XM_005272156.1 | Intron | XP_005272213.1 | ||||
XM_005272158.1 | Intron | XP_005272215.1 | ||||
XM_005272159.1 | Intron | XP_005272216.1 | ||||
XM_005272162.2 | Intron | XP_005272219.1 | ||||
XM_006716932.1 | Intron | XP_006716995.1 | ||||
XM_011518140.1 | Intron | XP_011516442.1 | ||||
XM_011518141.1 | Intron | XP_011516443.1 | ||||
XM_011518142.1 | Intron | XP_011516444.1 | ||||
XM_011518143.1 | Intron | XP_011516445.1 | ||||
XM_011518145.1 | Intron | XP_011516447.1 | ||||
XM_017014203.1 | Intron | XP_016869692.1 | ||||
XM_017014204.1 | Intron | XP_016869693.1 | ||||
XM_017014205.1 | Intron | XP_016869694.1 |
PRRC2B - proline rich coiled-coil 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |