Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118495 MIM: 601441 MIM: 162096 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRM4 - cholinergic receptor muscarinic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000741.3 | 1392 | Silent Mutation | ACC,ACT | T,T 447 | NP_000732.2 |
DGKZ - diacylglycerol kinase zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDK - midkine (neurite growth-promoting factor 2) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4688 - microRNA 4688 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |