Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCCACCCAAGCACGGCTGGGG[A/T]GAGGGGACAGGGTGCTGGCTTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCR1 - natural cytotoxicity triggering receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145457.2 | Intron | NP_001138929.2 | ||||
NM_001145458.2 | Intron | NP_001138930.2 | ||||
NM_001242356.2 | Intron | NP_001229285.1 | ||||
NM_001242357.2 | Intron | NP_001229286.1 | ||||
NM_004829.6 | Intron | NP_004820.2 | ||||
XM_011527528.2 | Intron | XP_011525830.1 | ||||
XM_011527529.2 | Intron | XP_011525831.1 | ||||
XM_011527530.2 | Intron | XP_011525832.1 |