Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTACCAAGCACAATGGCCACCAT[C/T]TGGGTTGCCTTCTTCTCCCGAAGTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 126451 | |||||||||||||||||||||||
Literature Links: |
DRD3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DRD3 - dopamine receptor D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000796.5 | 1410 | Silent Mutation | CAA,CAG | Q,Q 329 | NP_000787.2 | |
NM_001282563.2 | 1410 | Silent Mutation | CAA,CAG | Q,Q 329 | NP_001269492.1 | |
NM_001290809.1 | 1410 | Silent Mutation | CAA,CAG | Q,Q 329 | NP_001277738.1 | |
NM_033663.5 | 1410 | Silent Mutation | CAA,CAG | Q,Q 296 | NP_387512.3 | |
XM_017005829.1 | 1410 | Silent Mutation | CAA,CAG | Q,Q 329 | XP_016861318.1 |