Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAATCTTCTTTATCCCCTCACC[C/G]CCAGCACTATGGACCTGCAGGTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300369 MIM: 300574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACRC - acidic repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CXCR3 - C-X-C motif chemokine receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142797.1 | Intron | NP_001136269.1 | ||||
NM_001504.1 | Intron | NP_001495.1 | ||||
XM_005262256.3 | Intron | XP_005262313.1 | ||||
XM_005262257.3 | Intron | XP_005262314.1 | ||||
XM_017029435.1 | Intron | XP_016884924.1 | ||||
XM_017029436.1 | Intron | XP_016884925.1 |