Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C9orf24 - chromosome 9 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252195.1 | 767 | Missense Mutation | TAT,TGT | Y,C 114 | NP_001239124.1 | |
NM_032596.3 | 767 | Silent Mutation | GTA,GTG | V,V 247 | NP_115985.2 | |
NM_147168.1 | 767 | Silent Mutation | GTA,GTG | V,V 112 | NP_671697.1 | |
NM_147169.2 | 767 | Missense Mutation | TAT,TGT | Y,C 61 | NP_671698.1 | |
XM_005251616.4 | 767 | Missense Mutation | TAT,TGT | Y,C 249 | XP_005251673.1 | |
XM_005251618.4 | 767 | Missense Mutation | TAT,TGT | Y,C 114 | XP_005251675.1 | |
XM_011518055.2 | 767 | Missense Mutation | TAT,TGT | Y,C 114 | XP_011516357.1 |
KIAA1161 - KIAA1161 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |