Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605847 MIM: 600435 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL7C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BCL7C - BCL tumor suppressor 7C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CTF1 - cardiotrophin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142544.1 | 1456 | UTR 3 | NP_001136016.1 | |||
NM_001330.3 | 1456 | UTR 3 | NP_001321.1 | |||
XM_011545759.2 | 1456 | UTR 3 | XP_011544061.1 | |||
XM_011545760.2 | 1456 | UTR 3 | XP_011544062.1 |
MIR762 - microRNA 762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR762HG - MIR762 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |