Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAACCCCTACAACTTTCTAAG[A/T]TATGAGTAAATGATAGATGGAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603709 MIM: 182520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM22 - ADAM metallopeptidase domain 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRI - sorcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256891.1 | Intron | NP_001243820.1 | ||||
NM_001256892.1 | Intron | NP_001243821.1 | ||||
NM_003130.3 | Intron | NP_003121.1 | ||||
NM_198901.1 | Intron | NP_944490.1 | ||||
XM_011516528.1 | Intron | XP_011514830.1 |