Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGAGGATGAGTCAGAGAGCCA[A/G]GGTAGGTCCAGAAATCCAGGGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA1 - carbonic anhydrase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128829.3 | 666 | Missense Mutation | CCT,CTT | P,L 196 | NP_001122301.1 | |
NM_001128830.3 | 666 | Missense Mutation | CCT,CTT | P,L 196 | NP_001122302.1 | |
NM_001128831.3 | 666 | Missense Mutation | CCT,CTT | P,L 196 | NP_001122303.1 | |
NM_001164830.1 | 666 | Missense Mutation | CCT,CTT | P,L 196 | NP_001158302.1 | |
NM_001291967.1 | 666 | Missense Mutation | CCT,CTT | P,L 130 | NP_001278896.1 | |
NM_001291968.1 | 666 | Missense Mutation | CCT,CTT | P,L 83 | NP_001278897.1 | |
NM_001738.4 | 666 | Missense Mutation | CCT,CTT | P,L 196 | NP_001729.1 |