Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601663 MIM: 608442 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ESR2 - estrogen receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040275.1 | 3385 | UTR 3 | NP_001035365.1 | |||
NM_001214902.1 | 3385 | UTR 3 | NP_001201831.1 | |||
NM_001271876.1 | 3385 | Intron | NP_001258805.1 | |||
NM_001271877.1 | 3385 | Intron | NP_001258806.1 | |||
NM_001291712.1 | 3385 | UTR 3 | NP_001278641.1 | |||
NM_001291723.1 | 3385 | UTR 3 | NP_001278652.1 | |||
NM_001437.2 | 3385 | Intron | NP_001428.1 | |||
XM_017021079.1 | 3385 | Intron | XP_016876568.1 | |||
XM_017021080.1 | 3385 | Intron | XP_016876569.1 | |||
XM_017021081.1 | 3385 | Intron | XP_016876570.1 | |||
XM_017021082.1 | 3385 | Intron | XP_016876571.1 | |||
XM_017021083.1 | 3385 | Intron | XP_016876572.1 | |||
XM_017021084.1 | 3385 | Intron | XP_016876573.1 |