Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCTGTCCCGGTCATCCTTGAC[A/G]GTGTAATACTCCTTATCGGTTTCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147795 MIM: 609953 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
JAK1 - Janus kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320923.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001307852.1 | |
NM_001321852.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001308781.1 | |
NM_001321853.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001308782.1 | |
NM_001321854.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001308783.1 | |
NM_001321855.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001308784.1 | |
NM_001321856.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_001308785.1 | |
NM_001321857.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1035 | NP_001308786.1 | |
NM_002227.3 | 3419 | Silent Mutation | ACC,ACT | T,T 1036 | NP_002218.2 | |
XM_017001212.1 | 3419 | Silent Mutation | ACC,ACT | T,T 1035 | XP_016856701.1 |
RAVER2 - ribonucleoprotein, PTB binding 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |