Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGCGAGGCCCGGAGGGGGATG[C/G]GAACGTGAGTGTGGGCAAGGGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604159 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A5 - solute carrier family 6 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318369.1 | 668 | UTR 5 | NP_001305298.1 | |||
NM_004211.4 | 668 | Missense Mutation | GCG,GGG | A,G 132 | NP_004202.3 | |
XM_017018544.1 | 668 | Intron | XP_016874033.1 | |||
XM_017018545.1 | 668 | Intron | XP_016874034.1 |