Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603758 MIM: 607439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTZ1 - glutathione S-transferase zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312660.1 | 142 | Intron | NP_001299589.1 | |||
NM_145870.2 | 142 | Intron | NP_665877.1 | |||
NM_145871.2 | 142 | Intron | NP_665878.2 | |||
XM_011536670.2 | 142 | UTR 5 | XP_011534972.1 | |||
XM_011536671.2 | 142 | Intron | XP_011534973.1 | |||
XM_017021243.1 | 142 | Intron | XP_016876732.1 |
POMT2 - protein O-mannosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013382.5 | 142 | Intron | NP_037514.2 | |||
XM_011536675.2 | 142 | Intron | XP_011534977.1 | |||
XM_011536676.2 | 142 | Intron | XP_011534978.1 | |||
XM_011536677.2 | 142 | Intron | XP_011534979.1 | |||
XM_011536679.2 | 142 | Intron | XP_011534981.1 | |||
XM_017021244.1 | 142 | Intron | XP_016876733.1 | |||
XM_017021245.1 | 142 | Intron | XP_016876734.1 |