Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601998 MIM: 606583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRA - estrogen related receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282450.1 | 494 | Intron | NP_001269379.1 | |||
NM_001282451.1 | 494 | Intron | NP_001269380.1 | |||
NM_004451.4 | 494 | Intron | NP_004442.3 | |||
XM_017017313.1 | 494 | Intron | XP_016872802.1 |
PRDX5 - peroxiredoxin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012094.4 | 494 | Intron | NP_036226.1 | |||
NM_181651.2 | 494 | Intron | NP_857634.1 | |||
NM_181652.2 | 494 | Intron | NP_857635.1 | |||
XM_005273882.4 | 494 | Intron | XP_005273939.2 |
TRMT112 - tRNA methyltransferase 11-2 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286082.1 | 494 | UTR 5 | NP_001273011.1 | |||
NM_001286084.1 | 494 | UTR 5 | NP_001273013.1 | |||
NM_016404.2 | 494 | UTR 5 | NP_057488.1 |