Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTAACGCCTCTGCAGCGATTCCC[A/G]GTAGAAACGGAAGACGTGTTGCGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC5 - exosome component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020158.3 | 696 | Missense Mutation | CGG,TGG | R,W 225 | NP_064543.3 |
TMEM91 - transmembrane protein 91 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |