Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGAGAGAACATCAATGATAGCA[C/T]TGTAAAATCAAATGTTAATATTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615125 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD13C - ankyrin repeat domain 13C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030816.4 | Intron | NP_110443.3 | ||||
XM_005271234.3 | Intron | XP_005271291.1 | ||||
XM_005271235.3 | Intron | XP_005271292.1 | ||||
XM_006710929.3 | Intron | XP_006710992.1 | ||||
XM_017002413.1 | Intron | XP_016857902.1 | ||||
XM_017002414.1 | Intron | XP_016857903.1 | ||||
XM_017002415.1 | Intron | XP_016857904.1 | ||||
XM_017002416.1 | Intron | XP_016857905.1 |