Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGAGAAGCACACCTGTCCAGGG[A/C]CCACGATGTGTCTGGTGGTGTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL2RA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL2RA - interleukin 2 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000417.2 | Intron | NP_000408.1 | ||||
NM_001308242.1 | Intron | NP_001295171.1 | ||||
NM_001308243.1 | Intron | NP_001295172.1 |