Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCCAGCTCCTTTCCTCCCCTT[A/C]TTCCTCAATCAGGGATGCCAGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611848 MIM: 610348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR608 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR608 - microRNA 608 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL43 - mitochondrial ribosomal protein L43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4G - semaphorin 4G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203244.1 | Intron | NP_001190173.1 | ||||
NM_017893.3 | Intron | NP_060363.2 | ||||
XM_005270008.2 | Intron | XP_005270065.1 |
SLF2 - SMC5-SMC6 complex localization factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |