Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTGGGAAAAGATGGCGGCTGCC[C/G]CACAATCCCGGGTTGTCCGGGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604689 MIM: 603834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP3 - A-kinase anchoring protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278309.1 | 42 | Intron | NP_001265238.1 | |||
NM_006422.3 | 42 | Intron | NP_006413.3 | |||
XM_005253664.2 | 42 | Intron | XP_005253721.1 | |||
XM_011520908.1 | 42 | Intron | XP_011519210.1 | |||
XM_011520909.1 | 42 | Intron | XP_011519211.1 | |||
XM_011520910.2 | 42 | Intron | XP_011519212.1 |
NDUFA9 - NADH:ubiquinone oxidoreductase subunit A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005002.4 | 42 | Missense Mutation | CCA,GCA | P,A 5 | NP_004993.1 |