Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTTCTCCACTTTGAGTATACT[A/G]GCTAGATCACCAATATTACTTTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600367 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSTF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSTF3 - cleavage stimulation factor subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033505.1 | 1663 | Intron | NP_001028677.1 | |||
NM_001033506.1 | 1663 | Intron | NP_001028678.1 | |||
NM_001326.2 | 1663 | Silent Mutation | GCC,GCT | A,A 499 | NP_001317.1 |