Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCCACATCTAAATTCCATATTT[G/T]ATGTAATATAAGGAAAAATAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605988 MIM: 606503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLRE1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCLRE1C - DNA cross-link repair 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033855.2 | Intron | NP_001029027.1 | ||||
NM_001033857.2 | Intron | NP_001029029.1 | ||||
NM_001033858.2 | Intron | NP_001029030.1 | ||||
NM_001289076.1 | Intron | NP_001276005.1 | ||||
NM_001289077.1 | Intron | NP_001276006.1 | ||||
NM_001289078.1 | Intron | NP_001276007.1 | ||||
NM_001289079.1 | Intron | NP_001276008.1 | ||||
NM_022487.3 | Intron | NP_071932.2 |
SUV39H2 - suppressor of variegation 3-9 homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193424.1 | Intron | NP_001180353.1 | ||||
NM_001193425.1 | Intron | NP_001180354.1 | ||||
NM_001193426.1 | Intron | NP_001180355.1 | ||||
NM_001193427.1 | Intron | NP_001180356.1 | ||||
NM_024670.3 | Intron | NP_078946.1 | ||||
XM_006717503.3 | Intron | XP_006717566.1 | ||||
XM_011519662.2 | Intron | XP_011517964.1 | ||||
XM_017016637.1 | Intron | XP_016872126.1 |