Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCCAGTGAGTCCTAATGGTTT[C/T]ATTTCAGGCAGCAGATTCATTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609239 MIM: 614041 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPAR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPAR6 - lysophosphatidic acid receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162497.1 | 2664 | UTR 3 | NP_001155969.1 | |||
NM_001162498.1 | 2664 | UTR 3 | NP_001155970.1 | |||
NM_005767.5 | 2664 | UTR 3 | NP_005758.2 |
RB1 - RB transcriptional corepressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000321.2 | 2664 | Intron | NP_000312.2 | |||
XM_011535171.2 | 2664 | Intron | XP_011533473.1 |