Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGACACAGTGTCAACCGGGTCC[A/G]CGGTGAGTGGGGTGTTGTGGTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL2 - ADAMTS like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145320.1 | 303 | Missense Mutation | ACG,GCG | T,A 30 | NP_001138792.1 | |
NM_014694.3 | 303 | Missense Mutation | ACG,GCG | T,A 30 | NP_055509.2 | |
XM_005272237.3 | 303 | Missense Mutation | ACG,GCG | T,A 139 | XP_005272294.1 | |
XM_005272238.3 | 303 | Missense Mutation | CAC,CGC | H,R 89 | XP_005272295.1 | |
XM_005272239.2 | 303 | Missense Mutation | ACG,GCG | T,A 30 | XP_005272296.1 | |
XM_011519241.2 | 303 | Missense Mutation | ACG,GCG | T,A 139 | XP_011517543.2 | |
XM_011519242.2 | 303 | Missense Mutation | CAC,CGC | H,R 89 | XP_011517544.1 |