Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCAGCCGCGCGCAGGTACCGTG[C/A]GACATCGCGATGGCCCAGCTCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN2A - cyclin dependent kinase inhibitor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000077.4 | 709 | Missense Mutation | NP_000068.1 | |||
NM_001195132.1 | 709 | Missense Mutation | NP_001182061.1 | |||
NM_058195.3 | 709 | UTR 3 | NP_478102.2 | |||
NM_058197.4 | 709 | UTR 3 | NP_478104.2 | |||
XM_005251343.1 | 709 | Missense Mutation | XP_005251400.1 | |||
XM_011517675.2 | 709 | Missense Mutation | XP_011515977.1 | |||
XM_011517676.2 | 709 | Missense Mutation | XP_011515978.1 | |||
XM_011517679.1 | 709 | Missense Mutation | XP_011515981.1 |
CDKN2A-AS1 - CDKN2A antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |