Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGCTCTTCTGGGGGGCCCGGTA[C/T]GTCCGCACCGACTACACGCTGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSDMA - gasdermin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178171.4 | 277 | Silent Mutation | TAC,TAT | Y,Y 53 | NP_835465.2 | |
XM_006721832.2 | 277 | Silent Mutation | TAC,TAT | Y,Y 53 | XP_006721895.1 | |
XM_011524651.2 | 277 | Intron | XP_011522953.1 | |||
XM_017024502.1 | 277 | Silent Mutation | TAC,TAT | Y,Y 53 | XP_016879991.1 | |
XM_017024503.1 | 277 | Silent Mutation | TAC,TAT | Y,Y 53 | XP_016879992.1 | |
XM_017024504.1 | 277 | Silent Mutation | TAC,TAT | Y,Y 53 | XP_016879993.1 |