Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGTACGTCCGCACCGACTACAC[A/G]CTGCTGGATGTGCTTGAGCCCGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611218 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSDMA - gasdermin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178171.4 | 295 | Silent Mutation | ACA,ACG | T,T 59 | NP_835465.2 | |
XM_006721832.2 | 295 | Silent Mutation | ACA,ACG | T,T 59 | XP_006721895.1 | |
XM_011524651.2 | 295 | Intron | XP_011522953.1 | |||
XM_017024502.1 | 295 | Silent Mutation | ACA,ACG | T,T 59 | XP_016879991.1 | |
XM_017024503.1 | 295 | Silent Mutation | ACA,ACG | T,T 59 | XP_016879992.1 | |
XM_017024504.1 | 295 | Silent Mutation | ACA,ACG | T,T 59 | XP_016879993.1 |