Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611218 | ||||||||||||||||||||||||||||||||
Literature Links: |
GSDMA PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GSDMA - gasdermin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178171.4 | 500 | Missense Mutation | GTG,TTG | V,L 128 | NP_835465.2 | |
XM_006721832.2 | 500 | Missense Mutation | GTG,TTG | V,L 128 | XP_006721895.1 | |
XM_011524651.2 | 500 | Intron | XP_011522953.1 | |||
XM_017024502.1 | 500 | Missense Mutation | GTG,TTG | V,L 128 | XP_016879991.1 | |
XM_017024503.1 | 500 | Missense Mutation | GTG,TTG | V,L 128 | XP_016879992.1 | |
XM_017024504.1 | 500 | Missense Mutation | GTG,TTG | V,L 128 | XP_016879993.1 |