Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCTGTGTTATATACAAATCTTA[C/T]ACCTGCTCTTCTCCGTGACTCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC39 - leucine rich repeat containing 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256385.1 | Intron | NP_001243314.1 | ||||
NM_001256386.1 | Intron | NP_001243315.1 | ||||
NM_001256387.1 | Intron | NP_001243316.1 | ||||
NM_144620.3 | Intron | NP_653221.1 |
TRMT13 - tRNA methyltransferase 13 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |