Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGTATTAGAACATAGTATAAA[A/C]GTTAAAACAGCAGCTAGAATCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602374 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNN3 - calponin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286055.1 | Intron | NP_001272984.1 | ||||
NM_001286056.1 | Intron | NP_001272985.1 | ||||
NM_001839.4 | Intron | NP_001830.1 | ||||
XM_017000245.1 | Intron | XP_016855734.1 |
SLC44A3 - solute carrier family 44 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |