Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCGAATACCGCAGTAGCCACAG[C/T]GGCACCCACTCGTGCGCTTGAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611199 MIM: 601176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNTTIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNTTIP2 - deoxynucleotidyltransferase terminal interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCLM - glutamate-cysteine ligase modifier subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308253.1 | 1101 | Silent Mutation | CCA,CCG | P,P 222 | NP_001295182.1 | |
NM_002061.3 | 1101 | Silent Mutation | CCA,CCG | P,P 244 | NP_002052.1 | |
XM_011541261.2 | 1101 | Silent Mutation | CCA,CCG | P,P 156 | XP_011539563.1 | |
XM_017001056.1 | 1101 | UTR 3 | XP_016856545.1 | |||
XM_017001057.1 | 1101 | UTR 3 | XP_016856546.1 |