Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGACTACAGCGTGTGTTTTTTC[C/T]TTCCTCTTTCCCCTGCCTGTGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987013 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987013 - putative uncharacterized protein DDB_G0287265 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PALM2 - paralemmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037293.2 | 157 | Intron | NP_001032370.1 | |||
NM_053016.5 | 157 | UTR 5 | NP_443749.5 |
PALM2-AKAP2 - PALM2-AKAP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007203.4 | 157 | UTR 5 | NP_009134.1 | |||
NM_147150.2 | 157 | UTR 5 | NP_671492.1 |