Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTGGCAAGATGACTCCGTTATC[A/G]AATGAAAGAATGTTGTGGATTCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602894 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLRD1 - killer cell lectin like receptor D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114396.1 | Intron | NP_001107868.1 | ||||
NM_002262.3 | Intron | NP_002253.1 | ||||
NM_007334.2 | Intron | NP_031360.1 | ||||
XM_006719067.3 | Intron | XP_006719130.1 | ||||
XM_011520650.2 | Intron | XP_011518952.1 | ||||
XM_011520651.2 | Intron | XP_011518953.1 | ||||
XM_017019285.1 | Intron | XP_016874774.1 | ||||
XM_017019286.1 | Intron | XP_016874775.1 | ||||
XM_017019287.1 | Intron | XP_016874776.1 | ||||
XM_017019288.1 | Intron | XP_016874777.1 | ||||
XM_017019289.1 | Intron | XP_016874778.1 | ||||
XM_017019290.1 | Intron | XP_016874779.1 | ||||
XM_017019291.1 | Intron | XP_016874780.1 |
Set Membership: |
HapMap |