Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTGCTGGACCACACGGTAGCC[C/G]CCAGCCCATACTTAACGTTGTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606467 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH8A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALDH8A1 - aldehyde dehydrogenase 8 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193480.1 | 1178 | Missense Mutation | GCG,GGG | A,G 370 | NP_001180409.1 | |
NM_022568.3 | 1178 | Missense Mutation | GCG,GGG | A,G 420 | NP_072090.1 | |
NM_170771.2 | 1178 | Missense Mutation | GCG,GGG | A,G 366 | NP_739577.1 |
Set Membership: |
DME Validated Inventoried |