Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTAACTTCTCCAGCAATCAGAG[C/T]GCTCCCCCTCACATCAGTGGCATGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 179611 | |||||||||||||||||||||||
Literature Links: |
EPHA3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EPHA3 - EPH receptor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005233.5 | 82 | UTR 5 | NP_005224.2 | |||
NM_182644.2 | 82 | UTR 5 | NP_872585.1 | |||
XM_005264715.2 | 82 | UTR 5 | XP_005264772.1 | |||
XM_005264716.2 | 82 | UTR 5 | XP_005264773.1 |