Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGACTAAGAGCATGCATTCTATC[C/T]GAGGAGCTCAATGTCACACTGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606694 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP155 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NUP155 - nucleoporin 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278312.1 | 3863 | Silent Mutation | TCA,TCG | S,S 1174 | NP_001265241.1 | |
NM_004298.3 | 3863 | Silent Mutation | TCA,TCG | S,S 1179 | NP_004289.1 | |
NM_153485.2 | 3863 | Silent Mutation | TCA,TCG | S,S 1238 | NP_705618.1 | |
XM_011514165.2 | 3863 | Intron | XP_011512467.1 |
Set Membership: |
HapMap |