Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGACTTCCTGGTTTGCCAAGGCT[C/G]CCTCTTTGCTTGCCTCTGCGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS8 - HAUS augmin like complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011699.1 | 1088 | Missense Mutation | GCA,GGA | A,G 325 | NP_001011699.1 | |
NM_033417.1 | 1088 | Missense Mutation | GCA,GGA | A,G 326 | NP_219485.1 | |
XM_005260154.4 | 1088 | Missense Mutation | GCA,GGA | A,G 181 | XP_005260211.1 |