Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAAAGACTCGGCAGTCTGCAGC[A/G]GGGCGGGTGGGAGTAATTGTGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLK4 - polo like kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190799.1 | Intron | NP_001177728.1 | ||||
NM_001190801.1 | Intron | NP_001177730.1 | ||||
NM_014264.4 | Intron | NP_055079.3 | ||||
XM_005262701.2 | Intron | XP_005262758.1 | ||||
XM_017007662.1 | Intron | XP_016863151.1 | ||||
XM_017007663.1 | Intron | XP_016863152.1 |