Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCCCGCTTCCGCACTCCCAACA[C/T]GGGGAAGCTTCCAGCGCTGCCGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191039 MIM: 605574 | ||||||||||||||||||||
Literature Links: |
SNX21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX21 - sorting nexin family member 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNC2 - troponin C2, fast skeletal type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003279.2 | Intron | NP_003270.1 |
UBE2C - ubiquitin conjugating enzyme E2 C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |