Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGACAAAGAGAAGACTCTGCAG[C/G]AGACCTGCCCTGATACAAAGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA2 - annexin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002857.1 | Intron | NP_001002857.1 | ||||
NM_001002858.2 | Intron | NP_001002858.1 | ||||
NM_001136015.2 | Intron | NP_001129487.1 | ||||
NM_004039.2 | Intron | NP_004030.1 | ||||
XM_011521477.2 | Intron | XP_011519779.1 | ||||
XM_017022090.1 | Intron | XP_016877579.1 | ||||
XM_017022091.1 | Intron | XP_016877580.1 | ||||
XM_017022092.1 | Intron | XP_016877581.1 | ||||
XM_017022093.1 | Intron | XP_016877582.1 |