Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGTGGGCACCCACGTCTTCTC[A/G]CCCTGGACAAAGACCACTCCCCGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606682 MIM: 602254 MIM: 612747 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPS4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HPS4 - HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRRD - SRR1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013694.2 | 1485 | Intron | NP_001013716.2 | |||
XM_011530178.2 | 1485 | UTR 3 | XP_011528480.1 | |||
XM_017028799.1 | 1485 | Intron | XP_016884288.1 |
TFIP11 - tuftelin interacting protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008697.1 | 1485 | Silent Mutation | NP_001008697.1 | |||
NM_012143.2 | 1485 | Silent Mutation | NP_036275.1 | |||
XM_011530079.1 | 1485 | Silent Mutation | XP_011528381.1 | |||
XM_017028735.1 | 1485 | Silent Mutation | XP_016884224.1 |
Set Membership: |
HapMap |