Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAATCCAAGTCTTTGCTTATTTG[A/C]AATGCACAAACTATTTTTTTGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRB14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRB14 - growth factor receptor bound protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303422.1 | 1743 | UTR 3 | NP_001290351.1 | |||
NM_004490.2 | 1743 | UTR 3 | NP_004481.2 | |||
XM_011511022.2 | 1743 | Intron | XP_011509324.1 | |||
XM_017003899.1 | 1743 | UTR 3 | XP_016859388.1 |